ContactUsLinkComponent
Ice pan, solid, one piece molded, expanded polyvinyl chloride, unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 2.5l capacity, blue. 1 * 1 items
  BCISBCS-115-25B
 :  BIOCISION
Ice pan, solid, one piece molded, expanded polyvinyl chloride, unbreakable containment for variety of temperature media such as ice, dry ice, liquid nitrogen, saline solution, and acetone. 2.5l capacity, blue. 1 * 1 items
  BCISBCS-115-25B
 :  BIOCISION
Frequently Bought Together