Orders LinkContactUsLinkComponent
Write-on labels
Write-on labels
Write-on labels, Vinyl, Green on white, 58×25 mm, 225 labels/pack
Catalog # BRDY225084
Write-on labels
Catalog # BRDY225084
Supplier:  
Write-on labels
Write-on labels
Write-on labels, Vinyl, Green on white, 58×25 mm, 225 labels/pack
Discontinued
Product Details & Documents
These write-on labels provide easy identification for calibration, testing, and repair of tools and equipment.
  • Labels are used to track calibration, inspection, or parts and/or equipement
Write-on areas allow date, inspector, or other information to be periodically added.
Specifications
  • Colour:
    Green on white
  • Description:
    Write-on labels
  • Dimensions:
    58×25 mm
  • Material:
    Vinyl
  • Seq:
    17
  • Packed:
    225 labels/pack