Orders LinkContactUsLinkComponent
Gel drying Frames™
Gel drying Frames™
Cellophane, 140×140 mm
Catalog # DIVEAB-110762
Supplier:   DIVERSIFIED BIOTECH
Gel drying Frames™
Catalog # DIVEAB-110762
Gel drying Frames™
Gel drying Frames™
Cellophane, 140×140 mm
Price
44,70 €
Log in for availability
1 * 50 ST
Product Details & Documents
Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
Delivery information: The complete kit includes 2 open frames, 2 solid back plates, 12 side clips and 50 sheets of pre-cut cellophane.
Specifications
  • Description:
    Cellophane, 140×140 mm
  • Seq:
    02