Order Entry
Austria
Orders LinkContactUsLinkComponent
Plaster dispenser, QuickFix Detectable

Plaster dispenser, QuickFix Detectable

Plaster dispenser with 90 detectable plasters, 232×33×133 mm
Catalog # 118-0229
Supplier:   Plum Safety
Plaster dispenser, QuickFix Detectable
Catalog # 118-0229
Supplier:  Plum Safety
Plaster dispenser, QuickFix Detectable
Plaster dispenser, QuickFix Detectable

Plaster dispenser with 90 detectable plasters, 232×33×133 mm

Price
35,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dispenser containing highly visible blue plasters with an integrated metal surface that can be detected via a metal detector. The dispenser contains 90 plasters (2 refills) and can be wall-mounted.

  • Elastic textile fabric: breathable and skin friendly
  • Adapt to movements of the skin and are therefore very comfortable to wear
  • Very hygienic as every plaster is individually wrapped
  • Dispenser can be operated with just one hand
  • Easy to refill

Plaster size: 72×25 mm

Delivery information: Dispenser includes 2×45 plaster strips. A refill pack can be ordered separately.

Specifications
  • Description:
    Plaster dispenser with 90 detectable plasters
  • W×D×H:
    232×33×133 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.