Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 1 L
Catalog # CAYM600228-1
Supplier:   Cayman Chemical
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # CAYM600228-1
Supplier:  Cayman Chemical
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 1 L

CAS Number:  
Price
225,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.