Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 500 mL
Catalog # CAYM600215-500
Supplier: Cayman Chemical
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Catalog # CAYM600215-500
Supplier: Cayman Chemical
CAS Number:
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:EDTA buffer solution pH 8,0 (0,5 mol/l)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



