Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 ml
Catalog # CAYM600215-100
Supplier:   Cayman Chemical
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Catalog # CAYM600215-100
Supplier:  Cayman Chemical
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 ml

CAS Number:  
Price
34,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications