Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 8 (1 mol/L)

Buffer, TRIS solution pH 8 (1 mol/L)

TRIS buffer solution pH 8 (1 mol/L), Glass bottle, 250 ml
Catalog # CAYM600202-250
Supplier:   Cayman Chemical
Buffer, TRIS solution pH 8 (1 mol/L)
Catalog # CAYM600202-250
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TRIS solution pH 8 (1 mol/L)

TRIS buffer solution pH 8 (1 mol/L), Glass bottle, 250 ml

CAS Number:  
Price
64,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Description:
    TRIS buffer solution pH 8 (1 mol/L)

Specifications