Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (1 mol/L)

Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 500 mL
Catalog # CAYM600201-500
Supplier:   Cayman Chemical
Buffer, TRIS solution pH 7,5 (1 mol/L)
Catalog # CAYM600201-500
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
121,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TRIS buffer solution pH 7,5 (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.