Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (1 mol/L)

Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 250 ml
Catalog # CAYM600201-250
Supplier:   Cayman Chemical
Buffer, TRIS solution pH 7,5 (1 mol/L)
Catalog # CAYM600201-250
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 250 ml

CAS Number:  
Price
64,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Description:
    TRIS buffer solution pH 7,5 (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.