Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7 (1 mol/L)

Buffer, TRIS solution pH 7 (1 mol/L)

TRIS buffer solution pH 7 (1 mol/L), Glass bottle, 1 L
Catalog # CAYM600200-1
Supplier:   Cayman Chemical
Buffer, TRIS solution pH 7 (1 mol/L)
Catalog # CAYM600200-1
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TRIS solution pH 7 (1 mol/L)

TRIS buffer solution pH 7 (1 mol/L), Glass bottle, 1 L

CAS Number:  
Price
225,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS buffer solution pH 7 (1 mol/L)

Specifications