Estrone
Glass bottle for solids, 5 g
Catalog # CAYM10006485-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:53-16-7
- Formula:C₁₈H₂₂O₂
- MDL Number:MFCD00003620
- EINECS:200-164-5
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



