Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Estrone

Glass bottle for solids, 1 g
Artikelnr. CAYM10006485-1
Anbieter:   Cayman Chemical
Estrone
Artikelnr. CAYM10006485-1
Anbieter:  Cayman Chemical
CAS Nummer:  53-16-7
Estrone

Glass bottle for solids, 1 g

CAS Nummer:  53-16-7
Preis
29,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • MDL Number:MFCD00003620
  • EINECS:200-164-5

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications