ContactUsLinkComponent
Fridge-freezer,ctpsl2441 comfortgross capacity, refrigerator compartment overall [l] 187 gross capacity, freezer compartment [l] 53 energy ef ficiency class a++, star-coding 4, door hinges right interchangeable 1 * 1 items
Catalog # LIEBCTPSL2441
Supplier:  Liebherr
Fridge-freezer,ctpsl2441 comfortgross capacity, refrigerator compartment overall [l] 187 gross capacity, freezer compartment [l] 53 energy ef ficiency class a++, star-coding 4, door hinges right interchangeable 1 * 1 items
Catalog # LIEBCTPSL2441
Supplier:  Liebherr
Frequently Bought Together