ContactUsLinkComponent
ThermoScan DATALOGGER WITH CERTIFICATE comes with a temperature range from -40°C to +80°C and a resolution of 0.5°C. The Logger allows the documenting and checking of temperatures. The logger has a rewritable memory, a real-time clock and a fully fun 1 * 1 items
Catalog # DLTTD100002DE
Supplier:  DELTA T
ThermoScan DATALOGGER WITH CERTIFICATE comes with a temperature range from -40°C to +80°C and a resolution of 0.5°C. The Logger allows the documenting and checking of temperatures. The logger has a rewritable memory, a real-time clock and a fully fun 1 * 1 items
Catalog # DLTTD100002DE
Supplier:  DELTA T
Frequently Bought Together