Order Entry
Austria
Orders LinkContactUsLinkComponent
Funnels, mini

Funnels, mini

outlet Ø 10 mm, 50 mm, 31 mm
Catalog # BOCH8858
Supplier:   BOCHEM
Funnels, mini
Catalog # BOCH8858
Supplier:  BOCHEM
Funnels, mini
Funnels, mini

outlet Ø 10 mm, 50 mm, 31 mm

Price
4,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Mini funnel, made of 18/10 stainless steel.

  • Excellent temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • Funnel O-Ø:
    31 mm
  • Description:
    outlet Ø 10 mm
  • Length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.