ContactUsLinkComponent
Fitting for DIN support material d=26,9/12-13mm, cast iron/steel. Combines 26,9mm tubes with 12/13mm DIN rods. Consists in fitting (steel) and connecting workpiece (steel). Tube connector 26, rod and stainless steel tube are shown for demonstration 1 * 1 items
Catalog # BOCH111
Supplier:  BOCHEM
Fitting for DIN support material d=26,9/12-13mm, cast iron/steel. Combines 26,9mm tubes with 12/13mm DIN rods. Consists in fitting (steel) and connecting workpiece (steel). Tube connector 26, rod and stainless steel tube are shown for demonstration 1 * 1 items
Catalog # BOCH111
Supplier:  BOCHEM
Frequently Bought Together