Ready-to-use 1 mL and 5 mL pre-packed PRC columns. MEP, HEA and PPA HyperCel sorbents feature ligands to provide unique separation m echanisms different from conventional methods to capture or separate proteins. PRC COLUMN 8X100 MEP HYPERCEL, PRC08X 1 * 1 items
Catalog # 518-0089
Supplier: Cytiva (Formerly Pall Lab)
Frequently Bought Together