Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

9,21, Blue, 25 °C, 1 L
Catalog # METN51340230
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METN51340230
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions

9,21, Blue, 25 °C, 1 L

CAS Number:  
Price
60,39 €
Log in for availability
1 * 1 L
Product Details & Documents
  • For calibration of pH electrodes
  • Colour coded
  • Available as 1000 ml bottles

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH value:
    9,21

Specifications