ContactUsLinkComponent
19 COMPOSES DANS HNO3 5% Ag 100mg/l As 100mg/l Al 100mg/l B 100mg/l Ba 100mg/l Be 10mg/l Cd 10mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l Mn 100mg/l Ni 100mg/l Pb 100mg/l Sb 100mg/l Se 100mg/l Si 100mg/l V 100m 1 * 100 mL
Catalog # CMPL08F0.K1.5N.L1
19 COMPOSES DANS HNO3 5% Ag 100mg/l As 100mg/l Al 100mg/l B 100mg/l Ba 100mg/l Be 10mg/l Cd 10mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l Mn 100mg/l Ni 100mg/l Pb 100mg/l Sb 100mg/l Se 100mg/l Si 100mg/l V 100m 1 * 100 mL
Catalog # CMPL08F0.K1.5N.L1
Frequently Bought Together