V.P. TEST-REAGENT 10X10ML 1 * 1 KIT
Catalog # LIOF80280
Supplier: LIOFILCHEM
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
MICROBACT OXIDASE STRIPS 1 * 100 Tests
Supplier: THERMO FISHER DIAGNOSTICS