Order Entry
Austria
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

130 mm, 6 mm
Catalog # 231-0468
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0468
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro

130 mm, 6 mm

Price
36,50 €
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades

Documents

Specifications
  • Length:
    130 mm
  • Blade width:
    6 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.