Order Entry
Austria
Orders LinkContactUsLinkComponent
Double spatulas, micro | Avantor

Double spatulas, micro

210 mm, 4 mm
Artikelnr. 231-0465
Anbieter:   RSG
Double spatulas, micro
Artikelnr. 231-0465
Anbieter:  RSG
Double spatulas, micro
Double spatulas, micro

210 mm, 4 mm

Preis
37,60 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades

Documents

Spezifikationen
  • Length:
    210 mm
  • Blade width:
    4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.