Orders LinkContactUsLinkComponent
Test sets
Test sets
Test set I for fumes
Catalog # DRAE8103410
Supplier:   Dräger
Test sets
Catalog # DRAE8103410
Supplier:  Dräger
Test sets
Test sets
Test set I for fumes
Price
84,75 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
  • Description:
    Test set I for fumes
  • Seq:
    06
  • Cat. No.:
    DRAE8103410
  • Model:
    Test set I for fumes
  • Measurement time:
    180 s