Test sets
Test sets for fumes
Catalog # DRAE8103380
Supplier: Dräger
Jump to:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
- Bezeichnung:Test sets for fumes
- Seq:05
- Cat. No.:DRAE8103380
- Modell:Test sets for fumes
- Messdauer:240 s