Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Sacks

Storage Sacks

Approx. 1667 bags/pack, 0,05 mm, 200×300 mm
Catalog # LAMA116
Supplier:   LAMAPLAST
Storage Sacks
Catalog # LAMA116
Supplier:  LAMAPLAST
Storage Sacks
Storage Sacks

Approx. 1667 bags/pack, 0,05 mm, 200×300 mm

Price
260,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

LDPE, neutral storage bags.

  • With bottom weld
  • Can be used for foodstuffs
Specifications
  • Description:
    Approx. 1667 bags/pack
  • Dimensions:
    200×300 mm
  • Seq:
    02
  • Thickness:
    0,05 mm