Orders LinkContactUsLinkComponent
Pycnometers for bituminous materials
Pycnometers for bituminous materials
Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3, 1000 ml, 60
Catalog # NEUB0321-51-1000
Pycnometers for bituminous materials
Catalog # NEUB0321-51-1000
Pycnometers for bituminous materials
Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3, 1000 ml, 60
Price
264,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
  • Two-part assembly both parts with identification number
  • Cap with funnel for ease of filling
  • Ring mark on cap stem
Specifications
  • NS:
    60
  • Volume (nom.):
    1000 ml
  • Description:
    Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3
  • Seq:
    03