INSULIN (HUMAN) RECOMBINANT 1 * 8 mg
Catalog # EDQMI0310000
Supplier: EDQM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach