Order Entry
Austria
Orders LinkContactUsLinkComponent
Cooling systems, CoolSafe™

Cooling systems, CoolSafe™

CoolSafe™ chambers, aluminium, 384 well microplates, 384 in center, 10 on side (4 for 1,5 ml tubes/6 for o,5 ml tubes), 156×79×25 mm
Artikelnr. DIVECHAM-3840
Anbieter:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Artikelnr. DIVECHAM-3840
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™

CoolSafe™ chambers, aluminium, 384 well microplates, 384 in center, 10 on side (4 for 1,5 ml tubes/6 for o,5 ml tubes), 156×79×25 mm

Preis
1 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polystyrene cool safe box, aluminium cooling chamber.

  • Durable
  • Temperature stable
  • Corrosion resistant

Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.

Specifications
  • For:
    384 well microplates
  • No. of Holes:
    384 in center, 10 on side (4 for 1,5 ml tubes/6 for o,5 ml tubes)
  • Description:
    CoolSafe™ chambers, aluminium
  • Dimensions:
    156×79×25 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.