ChemiDoc-It 815 Cam/Dkrm Kit: 815 camera: 8.1 mp, 16-bit monochrome, digital camera and mount, with binning capabilities, cooling to -57C from ambient, MOTORISED fixed 30mm, f/1.4 lens, close-up diopter, USB 2.0 enhanced card and cables, Extremely k 1 * 1 items
Catalog # 733-2053
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together