0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
Orders Link
ContactUsLinkComponent
Buffers
pH Calibration Buffers
pH buffer solutions
pH buffer solutions
7,00, Green, 25 °C, 1 L
METN51340229
:
Mettler - Toledo
pH buffer solutions
METN51340229
:
Mettler - Toledo
:
pH buffer solutions
7,00, Green, 25 °C, 1 L
:
1 * 1 L
-
+
For calibration of pH electrodes
Colour coded
Available as 1000 ml bottles
Pk:
1 L
Reference temperature:
25 °C
Colour:
Green
pH value:
7,00