Brushes, FlexBrush™
Adapter, hook-shaped brush, twisted zinc with plastic handle, Polyester, Plastic, 292 mm
129-1068
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
- Bendable brush with polyester or microfibre cover
- Acme threaded end attaches to a variety of handles for extended reach
- Threaded handle (STA series) can be added for extended reach
- Covers are compatible with strong disinfectants and autoclavable
- Bristle material:Polyester
- Handle material:Plastic
- Description:Adapter, hook-shaped brush, twisted zinc with plastic handle
- Total length:292 mm
- Cat. No.:129-1068
- Environmentally Preferable:Waste Reducing