REAGENT REPLACEMENT PHOSPHATE 1 * 250 items
Catalog # PALIAP114
Supplier: Palintest
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH indicator test strips, PEHANON®
Supplier: MACHEREY-NAGEL
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, sulphite, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Ammonia Kit 0-0.1(N)Mg/L Pk50 With Calibrat Chart Palintest 1 * 50 items
Supplier: Palintest
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach