ContactUsLinkComponent
Graduated 0,3 mL Transfer Pipettes, Pediatric. Length 11,6 cm. Stem Diameter 3,0 mm. Overall Capacity 1,5 mL. Bulb Draw 0,8 mL. 22 drops per mL. 1 * 4.000 items
Catalog # SAMC282
Supplier:  SAMCO
Graduated 0,3 mL Transfer Pipettes, Pediatric. Length 11,6 cm. Stem Diameter 3,0 mm. Overall Capacity 1,5 mL. Bulb Draw 0,8 mL. 22 drops per mL. 1 * 4.000 items
Catalog # SAMC282
Supplier:  SAMCO