Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard (500 ml), 1413 µS, 1
Catalog # JENW16156-59
Supplier:   Cole-Parmer
Conductivity standards
Catalog # JENW16156-59
Supplier:  Cole-Parmer
CAS Number:  
Conductivity standards
Conductivity standard (500 ml), 1413 µS, 1
CAS Number:  
Price
83,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Standard solutions are used for calibrating conductivity cells.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Conductivity standard (500 ml)
  • Conductivity:
    1413 µS