Pipette Tips, Traditional
Product Details & Documents
These thin-wall tips are designed to fit RAININ and Pipetman pipettes with conical shafts, Eppendorf and Labsystems pipettes. Some tips feature a hydrophobic filter which forms a barrier against aerosols and liquids eliminating potential sample and pipette cross contamination.
- Filtered and pre-sterilised bioclean tips are gamma-irradiated
- ShaftGard tips are longer than standard tips for easy access into microcentrifuge and PCR tubes
- Wide orifice tips minimise shear forces on delicate samples and flow resistance for viscous samples
The tips are available in different packaging options:
Racks with removable cover
- Sealed bottoms and overlapping lids protect tips from dust, dirt and micro-organisms
- Two-position lid allows autoclaving with the lid in vent position
- Sturdy shelf construction allows easy tip loading with single and multi-channel pipettes
Green-Pak® Individual Refills and Green-Pak® SpaceSaver® Refills
- Environmentally friendly refills for racks; less plastic waste
- Individual refills contain 96 tips in 8×12 format, sealed in a shell of Code 1 PET recyclable plastic
- Each refill is completely sealed to protect the tips; hands and fingers never come in contact with tips during loading
- SpaceSaver® systems are provided with either eight or ten stacked refills
- SpaceSaver® refills are packaged in a code 1 PET sleeve which functions as an automatic tip dispenser that aligns and refills an empty rack with 96 tips at a time
Bulk
- Packaged in zip-lock bags
- Very economical
Documents
Specifications
- Packed:10 racks, 96 each
- Sterile:-
- Volume:10 µl
- Seq:13
- Tip style:Standard
- For use with:RAININ®, Pipetman® and other conical shaft pipettes, 2 and 10 μl