Order Entry
Austria
Orders LinkContactUsLinkComponent
 

inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench

inoLab® Multi 9310 Set 2, instrument with digital IDS pH electrode SenTix® 980
  665-0251
 :   WTW
inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench
  665-0251
 :  WTW
inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench
inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench

inoLab® Multi 9310 Set 2, instrument with digital IDS pH electrode SenTix® 980

 
1 860,00 €
   
1 * 1 ST
 

The inoLab® Multi 9310 is ideal for all laboratories that require complete documentation, or want to have the most efficient, error-free measurement processes. For the measurement of a single parameter from pH, conductivity and DO.

  • Continuous Measurement Control (CMC) for measuring in the optimum pH range
  • Digital sensor recognition
  • Quality Sensor Control (QSC) benchmarks the current condition of the special IDS pH electrodes
  • Digital processing and data transfer (GLP compliant) via USB interface
  • Optional built-in printer for printing of all measurement and calibration results
  • 'Wireless ready': Can be used with the new wireless IDS plug head sensors (wireless modules are required, not included in the delivery)

Intelligent digital sensors (IDS) provide sophisticated electrochemistry measuring systems in a single meter. The inolab® Multi 9310 IDS measures pH, mV, DO (saturation, concentration, partial pressure), conductivity (specific resistance, salinity, TDS), temperature and turbidity.

 : Supplied with universal power supply, stand, CD-ROM including software and USB cable. The application sets also include sensors.

 
  • Description:
    inoLab® Multi 9310 Set 2, instrument with digital IDS pH electrode SenTix® 980
  • Seq:
    03
  • Cat. No.:
    665-0251
  • Data logging - log function:
    Manual/timer controlled
  • pH range:
    Depending on sensor
  • Model:
    Multi 9310 IDS
  • Conductivity range:
    Depending on sensor
  • Outputs:
    Mini USB-B
  • Data logging capacity:
    Manual: 500 data sets; automatic: 4500 data sets
  • Power supply:
    Universal power supply or batteries (4× 1,5 V AA or 4× 1,2 V NiMH)
  • DO concentration:
    Depending on sensor
  • DO saturation:
    Depending on sensor
  • Display:
    Graphic LCD, backlit
Accessories

 

  •  
    Description
    Type
    Concentration
     
     
  • 664-0050
    IDS dissolved oxygen probe FDO® 925
    FDO® 925
    0,00 - 20,00 mg/L
       
    1 * 1 ST - 1 120,00 €

 

  •  
    Description
    Type
    Conductivity
     
     
  • 663-0152
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
    TetraCon® 925, 4-cell graphite
    10 µS/cm - 2000 mS/cm
       
    1 * 1 ST - 639,00 €

 

  •  
    Description
    Type
    pH
     
     
  • 662-1254
    IDS pH electrode Sentix® 980
    Sentix® 980
    0,000 - 14,000 (±0,004)
       
    1 * 1 ST - 332,00 €

 

  •  
    Description
    Type
    pH
     
     
  • 662-1249
    IDS pH electrode Sentix® 940
    Sentix® 940
    0,000 - 14,000 (±0,004)
       
    1 * 1 ST - 294,00 €

 

  •  
    Description
    Type
    Connection
     
     
  • 662-2020
    ORP electrode ORP-T 900
    ORP-T 900
    IDS plug, 1,5 m cable
       
    1 * 1 ST - 305,00 €

 

  •  
    Description
     
     
  • WTWA250049
    Thermal printer with paper P3002
       
    1 * 1 ST - 873,00 €