Order Entry
Austria
Orders LinkContactUsLinkComponent
Coloured cap inserts

Coloured cap inserts

Yellow
Catalog # 525-0501
Supplier:   Brand
Coloured cap inserts
Catalog # 525-0501
Supplier:  Brand
Coloured cap inserts
Coloured cap inserts

Yellow

Price
20,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

PP.

  • For quick identification
  • For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
  • Applications: −196 to +121 °C
Specifications
  • Colour:
    Yellow
  • Seq:
    05