Orders LinkContactUsLinkComponent
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
PVDF, 0,2 µm, Red
Catalog # 548-1404
Supplier:   THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Catalog # 548-1404
Supplier:  THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
PVDF, 0,2 µm, Red
Price
1 860,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
  • Compatible with both UHPLC and standard HPLC autosamplers
  • Colour coded pre-slit caps for easy identification of membrane and porosity
  • Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Specifications
  • Membrane:
    PVDF
  • Pore size:
    0,2 µm
  • Colour:
    Red
  • Seq:
    08