STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST38 STAINLESS STEEL TANK, 38L CAPACITY, INCL DRAIN TAP AN D PUMP. TEMP RANGE AMBIENT+5 TO 150C. 1 * 1 items
Catalog # GRANTX150-ST38EU
Supplier: GRANT INSTRUMENTS
Frequently Bought Together