Orders LinkContactUsLinkComponent
Micro-fluidic platform, SilFlow™
Micro-fluidic platform, SilFlow™
GC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm ID
Catalog # 548-1786
Micro-fluidic platform, SilFlow™
Catalog # 548-1786
Micro-fluidic platform, SilFlow™
Micro-fluidic platform, SilFlow™
GC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm ID
Price
842,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
SilFlow™ microchannel device (MCD) options for three-port GC splitters, four-port GC splitters and a Deans' switch MCD, perfect for multi-dimensional analyses. SilFlow™ can be incorporated into the system without impacting the chromatography.
  • Chemically-deactivated stainless steel channels avoiding active sites experienced with conventional connections
  • Conventional graphite or Vespel® ferrules cannot be used with the SilFlow™ MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection, giving optimised peak shapes
  • Can be operated at pressures greater than 25000 psi
  • SilFlow™ tracks oven temperature up to 20 °C/min. The design of SilFlow™ alleviates cold spots and sample condensation
 : SilFlow™ kits contain: SilFlow™ microchannel device (MCD), SilTite™ FingerTite tool, mounting bracket, appropriately sized ferrules, nuts and blanking ferrules for set up. 1,1 mm Ø tubing is not supplied in the splitter kits and must be ordered separately.
Documents
Specifications
  • Description:
    GC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm ID
  • Seq:
    08