Orders LinkContactUsLinkComponent
Stopcocks for dessicator lids
Stopcocks for dessicator lids
Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
Catalog # WITG0.246.002
Supplier:   witeg Labortechnik
Stopcocks for dessicator lids
Catalog # WITG0.246.002
Supplier:  witeg Labortechnik
Stopcocks for dessicator lids
Stopcocks for dessicator lids
Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
Price
41,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
  • Seq:
    019