Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Wire baskets, rectangular, with lid

220×140×50 mm
  HUPF7501133
Wire baskets, rectangular, with lid
  HUPF7501133
Wire baskets, rectangular, with lid
Wire baskets, rectangular, with lid

220×140×50 mm

 
384,00 €
   
1 * 1 ST
 

These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.

  • With lid
  • With base frame
  • Mesh size: 1,6×1,6 mm

Colour: Silver.

 
  • Seq:
    05
  • W×D×H:
    220×140×50 mm