ContactUsLinkComponent
Jetpei-macrophage is a dna transfection reagent dedicated to the transfection of macrophage primary cells and cell lines which expre ss mannose receptors. 1 * 100 µl
Catalog # 103-01N
undefined
Jetpei-macrophage is a dna transfection reagent dedicated to the transfection of macrophage primary cells and cell lines which expre ss mannose receptors. 1 * 100 µl
Catalog # 103-01N