ContactUsLinkComponent
The Polycarbonate Carboy is made from a clear material, heat stable and is appropriate for most life science solutions. It has easy- to-read graduation marks. Handles are at the top and bottom for easy lifting, filling, and pouring. The cap provides 1 * 1 items
Catalog # 216-0620
Supplier:  VWR Collection
undefined
The Polycarbonate Carboy is made from a clear material, heat stable and is appropriate for most life science solutions. It has easy- to-read graduation marks. Handles are at the top and bottom for easy lifting, filling, and pouring. The cap provides 1 * 1 items
Catalog # 216-0620
Supplier:  VWR Collection
Frequently Bought Together