Order Entry
Austria
Orders LinkContactUsLinkComponent
Drum stands, FB | Avantor

Drum stands, FB

Drum stands, FB 1, fire-galvanised, capacity 1×200 l drum, 600×650×415 mm
Artikelnr. LACOP21-1701-A
Drum stands, FB
Artikelnr. LACOP21-1701-A
Drum stands, FB

Drum stands, FB 1, fire-galvanised, capacity 1×200 l drum, 600×650×415 mm

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.

These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.

Spezifikationen
  • Description:
    Drum stands, FB 1, fire-galvanised, capacity 1×200 l drum
  • Seq:
    01
  • W×D×H:
    600×650×415 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.