Order Entry
Austria
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol, Sigma-Aldrich®

5 kg
Catalog # SIAL63559-5KG
Supplier:   Merck
D-(-)-Mannitol, Sigma-Aldrich®
Catalog # SIAL63559-5KG
Supplier:  Merck
CAS Number:  69-65-8
D-(-)-Mannitol, Sigma-Aldrich®
D-(-)-Mannitol, Sigma-Aldrich®

5 kg

CAS Number:  69-65-8
Price
903,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pk:
    5 kg

Specifications