Pycnometers for bituminous materials
Pycnometer with ring mark, matt shield and numbering, soda galss, 1000 ml, 60
NEUB0321-50-1000
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
- NS:60
- Volume (nom.):1000 ml
- Description:Pycnometer with ring mark, matt shield and numbering, soda galss
- Seq:01