Storage tubes, barcoded, Corning®
96× 1D/2D barcoded storage tubes, with screw caps, 1,3 ml, Orange, racked with cover, do 1,1 ml
Supplier: Corning
Jump to:
Product Details & Documents
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
- Maximum information, 14×14 dot 2D barcode
- Barcode stability, laser-etched, not an attached label
- Compound compatibility, polypropylene construction throughout, making it inert
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:racked with cover
- Recommended working volume:do 1,1 ml
- Capacity:1,3 ml
- Colour:Orange
- Description:96× 1D/2D barcoded storage tubes, with screw caps
Accessories
Product Information
- Catalog #ColourPackedAvailabilityPrice
- Empty racks with lids for 384 round or square storage tubesEmpty racks with lids for 384 round or square storage tubes731-0248Blue-Log in for availability1 * 20 ST - 219,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- 384× storage tube plug cap on mats for round tube, 384 caps per mat384× storage tube plug cap on mats for round tube, 384 caps per mat731-0247Orange384 caps per matLog in for availability1 * 38.400 ST - 4 260,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- 384× storage tube plug cap for round tube384× storage tube plug cap for round tube731-0246Orangebulk packLog in for availability1 * 38.400 ST - 3 040,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- Empty racks with lids for 96 screw capped or Thermoplastic Elastomers (TPE) capped storage tubesEmpty racks with lids for 96 screw capped or Thermoplastic Elastomers (TPE) capped storage tubes731-0245Natural-Log in for availability1 * 10 ST - 81,10 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum capThermoplastic elastomer (TPE) 96 storage tube septum cap731-0243Whitebulk packedLog in for availability1 * 9.600 ST - 590,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum cap on matThermoplastic elastomer (TPE) 96 storage tube septum cap on mat731-0244White96 caps per matLog in for availability1 * 4.800 ST - 826,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- 96× storage tube screw cap, with O-Ring, on mat, clear with cover96× storage tube screw cap, with O-Ring, on mat, clear with cover731-0242Orange96 caps per matLog in for availability1 * 4.800 ST - 3 400,00 €
Product Information
- Catalog #ColourPackedAvailabilityPrice
- 96× screw caps for storage tubes 1,3 ml, with O-Ring96× screw caps for storage tubes 1,3 ml, with O-Ring731-0241Orangebulk packedLog in for availability1 * 4.800 ST - 2 830,00 €



