Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage tubes, barcoded, Corning®

Storage tubes, barcoded, Corning®

96× 1D/2D barcoded storage tubes, without screw caps, 1,3 ml, Natural, racked with cover, do 1,1 ml
Catalog # 479-0969
Supplier:   Corning
Storage tubes, barcoded, Corning®
Catalog # 479-0969
Supplier:  Corning
Storage tubes, barcoded, Corning®
Storage tubes, barcoded, Corning®

96× 1D/2D barcoded storage tubes, without screw caps, 1,3 ml, Natural, racked with cover, do 1,1 ml

Price
973,00 €
Log in for availability
1 * 960 ST
Product Details & Documents

PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.

  • Maximum information, 14×14 dot 2D barcode
  • Barcode stability, laser-etched, not an attached label
  • Compound compatibility, polypropylene construction throughout, making it inert

Documents

Specifications
  • Packed:
    racked with cover
  • Recommended working volume:
    do 1,1 ml
  • Capacity:
    1,3 ml
  • Colour:
    Natural
  • Description:
    96× 1D/2D barcoded storage tubes, without screw caps
Accessories

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • Empty racks with lids for 384 round or square storage tubes
    Empty racks with lids for 384 round or square storage tubes
    731-0248
    Blue
    -
    Log in for availability
    1 * 20 ST - 219,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • 384× storage tube plug cap on mats for round tube, 384 caps per mat
    384× storage tube plug cap on mats for round tube, 384 caps per mat
    731-0247
    Orange
    384 caps per mat
    Log in for availability
    1 * 38.400 ST - 4 260,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • 384× storage tube plug cap for round tube
    384× storage tube plug cap for round tube
    731-0246
    Orange
    bulk pack
    Log in for availability
    1 * 38.400 ST - 3 040,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • Empty racks with lids for 96 screw capped or Thermoplastic Elastomers (TPE) capped storage tubes
    Empty racks with lids for 96 screw capped or Thermoplastic Elastomers (TPE) capped storage tubes
    731-0245
    Natural
    -
    Log in for availability
    1 * 10 ST - 81,10 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • Thermoplastic elastomer (TPE) 96 storage tube septum cap
    Thermoplastic elastomer (TPE) 96 storage tube septum cap
    731-0243
    White
    bulk packed
    Log in for availability
    1 * 9.600 ST - 590,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat
    Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat
    731-0244
    White
    96 caps per mat
    Log in for availability
    1 * 4.800 ST - 826,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • 96× storage tube screw cap, with O-Ring, on mat, clear with cover
    96× storage tube screw cap, with O-Ring, on mat, clear with cover
    731-0242
    Orange
    96 caps per mat
    Log in for availability
    1 * 4.800 ST - 3 400,00 €

Product Information

  • Catalog #
    Colour
    Packed
    Availability
    Price
  • 96× screw caps for storage tubes 1,3 ml, with O-Ring
    96× screw caps for storage tubes 1,3 ml, with O-Ring
    731-0241
    Orange
    bulk packed
    Log in for availability
    1 * 4.800 ST - 2 830,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.